missing translation for 'onlineSavingsMsg'
Learn More

VEE GEE Alcohol Hydrometer

Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.

Brand:  VEE GEE 6613-N

 View more versions of this product

Product Code. 17806573

  • 75.00€ / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Alcohol proof range differ depending on model
  • IRS Specification Size differ depending on model
  • li>Calibration for use at 15.5°C/60°F
  • Approx. Length: 305 mm
Specifications

Specifications

Alcohol Hydrometer
30.5 cm
Glass
125% to 145%
0.2%/±0.3%
Alcohol Hydrometer, Proof Scale, IRS Specification, Size N
15.5°C (60°F)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers