Learn More
VEE GEE Alcohol Hydrometer
Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.
Brand:  VEE GEE 6613-I
 View more versions of this product
Product Code. 17896553
  • 75.00€ / Each
Estimated Shipment: 06-01-2025
Description
  • Alcohol proof range differ depending on model
  • IRS Specification Size differ depending on model
  • li>Calibration for use at 15.5°C/60°F
  • Approx. Length: 305 mm
Specifications
Specifications
Alcohol Hydrometer
30.5 cm
Glass
60 to 80%
0.2%/±0.3%
Alcohol Hydrometer, Proof Scale, IRS Specification, Size I
15.5°C (60°F)