Learn More
VEE GEE Alcohol Hydrometer with Certification
Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.
475.00€
Specifications
Type | Alcohol Hydrometer |
---|---|
Length (Metric) | 30.5 cm |
Temperature of Standardization | 15.5°C (60°F) |
Product Code | Brand | Alcohol Range | Increments | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Alcohol Range | Increments | Price | Quantity & Availability | ||||
17876563
|
VEE GEE
6613-H-C |
40 to 60% | 0.20% |
475.00€
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
17816573
|
VEE GEE
6613-P-C |
145 to 165% | 0.20% |
475.00€
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
17816563
|
VEE GEE
6613-F-C |
0 to 20% | 0.20% |
475.00€
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
17806563
|
VEE GEE
6613-B-C |
80 to 120% | 0.50% |
475.00€
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
17876523
|
VEE GEE
6613-I-C |
60 to 80% | 0.50% |
475.00€
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
17846473
|
VEE GEE
6613-L-C |
90 to 110% | 0.20% |
475.00€
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
17856473
|
VEE GEE
6613-G-C |
20 to 40% | 0.20% |
475.00€
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
17896473
|
VEE GEE
6613-M-C |
105 to 125% | 0.20% |
475.00€
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
17876443
|
VEE GEE
6613-N-C |
125 to 145% | 0.20% |
475.00€
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
17826483
|
VEE GEE
6613-K-C |
75 to 95% | 0.20% |
475.00€
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
17896483
|
VEE GEE
6613-Q-C |
165 to 185% | 0.20% |
475.00€
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
17886483
|
VEE GEE
6613-R-C |
185 to 206% | 0.20% |
475.00€
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||||
Description
- Alcohol proof range differ depending on model
- IRS Specification Size differ depending on model li>Calibration for use at 15.5°C/60°F
- Approx. Length: 305 mm
Specifications
Alcohol Hydrometer | |
30.5 cm | |
15.5°C (60°F) |