Learn More
Alkaline running buffer soln., Thermo Scientific™
26.80€ - 52.36€
Specifications
Physical Form Liquid
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15435439
View Documents
Thermo Scientific Alfa Aesar
J63170.AK
250 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
15445439
View Documents
Thermo Scientific Alfa Aesar
J63170.AP
500 mL
52.36€
500mL
Estimated Shipment: 19-12-2024
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Liquid
Videos
Safety and Handling
Safety and Handling
Recommended Storage : Ambient temperatures