missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Allopregnanolone ELISA Kit

The Allopregnanolone Competitive ELISA quantitates allopregnanolone in serum, plasma, urine, saliva, dried fecal extracts or cell culture medium.

Brand:  Invitrogen™ EIAALLOX10

 View more versions of this product

Product Code. 17815117

  • 4635.00€ / Pack of 10
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Allopregnanolone Competitive ELISA quantitates allopregnanolone in serum, plasma, urine, saliva, dried fecal extracts or cell culture medium.∣Principle of the method: The Progesterone Metabolites Competitive ELISA research-use-only kit is designed to quantitatively measure allopregnanolone independent of species. A allopregnanolone standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit allopregnanolone antibodies and allopregnanolone-peroxidase conjugate is added to the wells. The allopregnanolone-peroxidase conjugate and any allopregnanolone in the sample will compete to bind to the anti-rabbit allopregnanolone antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound allopregnanolone-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of allopregnanolone present in the sample.∣Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3 alpha,5 alpha-tetrahydroprogesterone; 3 alpha-hydroxy-5 alpha-pregnan-20-one; allopregnanolone; Brexanolone; C21H34O2; Zulresso
129.7 pg/mL
ELISA
Colorimetric Microplate Reader
7%
HRP
Serum (50 μL), Plasma (50 μL), Fecal Extract (50 μL), Urine (12.5 μL), Supernatant (50 μL)
Allopregnanolone
16 hr. 45 min.
156.25 to 10,000 pg/mL
HRP
10 x 96 Assays
8%
Anti-IgG coated 96 well plate
Standard
Aassay Buffer Concentrate
Capture Antibody
HRP-conjugated Antigen
Wash Buffer Concentrate
TMB Substrate
Stop Solution
Plate Sealer
RUO
-20°C
45 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers