missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec 3000 Coverall, Hood
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Designed to protect, typical innovative features include dual zip systems and double cuffs
- Lightweight and durable
- Highly visible bright yellow color for improved worker safety
- Anti-static
Specifications
Specifications
Size | 3X-Large |
Material | Multi-layer Barrier Laminate Fabric |
Color | Yellow |
Seam Type | Welded |
Certifications/Compliance (Europe) | EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003 |
Model | Alphatec Model 121 |
For Use With (Application) | Transferring Liquids and Solids Between Vessels and Tanks and Process Equipment, Unexpected Leakages Spills or Other Releases, Opening and Draining Pumps Valves or Lines, Loading and Unloading Trucks and Vehicles, Filling, Blending and Charging of Raw Materials, Product Inspection, Mixing or Loading Chemicals, Cleaning of Plant and Machines |
Weight (Metric) | 80 gsm |
Style | Y30W121 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'