Learn More
Ansell AlphaTec™ 3000 Overboots Ultrasonically Welded Model 406
Shop All Ansell Products
Click to view available options
Size: 42 to 46
Description
- One of the lightest and most comfortable chemical protective garments on the market today, this durable multi-layer fabric provides an extremely effective barrier against both inorganic chemicals and biological hazards.
- Provides a strong liquid and particle barrier
- Overboots, elastic to top of boot, tie fastening and welded seams.
Specifications
Specifications
Size | 42 to 46 |
Material | Multi-layer Barrier Laminate Fabric |
Color | Yellow |
Disposable | Reusable |
Sterility | Non-sterile |
Seam Type | Welded |
For Use With (Application) | Unexpected leakages spills or other releases, Product inspection, Cleaning of plant and machines, Transferring liquids and solids between vessels and tanks and process equipment, Opening and draining pumps valves or lines, Loading and unloading of trucs and vehicles, Filling, blending and charging of raw materials, Mixing or loading chemicals |
Certifications/Compliance | CE Number, EN 14126:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN 13034:2005 + A1:2009 |
Resistance | Chemical-resistant, Biological Hazards |
Product Type | Boot Covers |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Ansell AlphaTec™ 3000 Overboots Ultrasonically Welded Model 406 >