Learn More
Ansell AlphaTec™ 38-514 Chemical Resistant Butyl Gloves Product Code.: 11721693
Only undefined left
Add to basket
Ansell AlphaTec™ 38-514 Chemical Resistant Butyl Gloves
Product Code. 11721693 Shop All Ansell Products
7
Click to view available options
Size:
7
8
9
10
11
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11721693
Brand: Ansell 38514070
Only undefined left
Add to basket
Only undefined left
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Thin butyl glove provides resistance to aggressive chemicals including solvents and strong acids.
  • Protection against aldehydes, ketones and esters as well as concentrated mineral acids
  • Flexible and comfortable
  • Natural, curved ergonomic shape
  • Easy donning and secure grip
  • Rough finish offers safer handling
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 3
Certifications/Compliance EN 374, EN 388, EN 1149
Certifications/Compliance (Europe) CE 0493, EN 1149, EN 16350, EN 420:2003 + A1:2009; Category III
Chlorinated No
Applications Industrial
Coating Material Butyl Polymer
Color Black
Latex Free Yes
Material Butyl
Powdered Powder Free
Product Type Butyl Gloves
Size 7
Thickness (English) 14 mil
Thickness (Metric) 0.35 mm
EN388 Standard EN 388:1000X, EN 388:1000
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ABCIKMPST
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Chemical plants and transportation, chemical processing, handling acids, alkalies, salts and ketones
Glove Cuff Style Beaded
Grip Design Rough
Hem Color Black
Lining Unlined
Model 38-514_7
Packaging Type Case
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Days
    25
    Hours
    11
    Minutes
    42
    Seconds
    29
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Ansell AlphaTec™ 38-514 Chemical Resistant Butyl Gloves >