Learn More
Fisherbrand™ Scaffolding Rods
Provides for sturdy scaffolding construction in the laboratory. Fisherbrand™ Scaffolding Rods are manufactured from a durable aluminum alloy.
Brand:  Fisherbrand™ 2000/3
 View more versions of this product
Product Code. 11577023
  • 14.85€ / Each
Estimated Shipment: 03-06-2025
Description
  • Approx. 13mm in diameter
  • Available in a range of lengths
  • Fits with Fisherbrand scaffolding connectors and foot adapters
Specifications
Specifications
Aluminum Alloy
1200 mm
Rod
Certificates
View accessory products
Product Title
Fisherbrand™ Scaffolding Rods > Length: 1200mm