Learn More
Rogo Sampaic™ Aluminum Lab Jack
Brand:  Rogo Sampaic™ FCD002
 View more versions of this product
Product Code. 11747265
  • 176.00€ / Each
Estimated Shipment: 04-06-2025
Specifications
Anodized Aluminum
65 to 280 mm
Lab Jack
1 Ea.
Product Title
Rogo Sampaic™ Aluminum Lab Jack > Height: 65 to 280mm