Learn More
Rogo Sampaic™ Aluminum Lab Jack
Brand:  Rogo Sampaic™ FCD003
 View more versions of this product
Product Code. 11757265
  • 261.00€ / Each
In Stock
Edge
Specifications
Anodized Aluminum
65 to 290 mm
Lab Jack
1 Ea.
Product Title
Rogo Sampaic™ Aluminum Lab Jack > Height: 65 to 290mm