Learn More
Gravis Aluminum Open Caps
Click to view available options
Material:
Aluminum
Description
- 40mm in diameter
- Autoclavable
Specifications
Material | Aluminum |
No. per Pack | 100Pack |
Quantity | 100 Pack |
Product Type | Open Top Cap |
Closure Material | Aluminum |
Closure Type | Thread 40 mm |