Learn More
Korff™ Aluminum Professional Rolls
46.50€ - 142.00€
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | ||||||
11946046
|
Korff™
60100 |
300mm | N/A | |||||||
10533282
|
Korff™
60100 |
300mm |
106.00€
150m |
In Stock
Log in to see stock. ![]() |
||||||
10322232
|
Korff™
60150 |
600mm |
142.00€
100m |
In Stock
Log in to see stock. ![]() |
||||||
10730193
|
Korff™
60110 |
450mm |
78.15€
150m |
Estimated Shipment: 20-06-2025
Log in to see stock. |
||||||
Specifications
Professional Rolls | |
Aluminum |
Product Title
Korff™ Aluminum Professional Rolls