Learn More
Korff™ Aluminum Sheets
61.10€ - 135.00€
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | ||||||
11972563
|
Korff™
60285 |
150mm |
135.00€
Pack of 1000 |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
10372322
|
Korff™
60270 |
80mm |
61.10€
Pack of 1000 |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
Specifications
Sheets | |
Aluminum |
Product Title
Korff™ Aluminum Sheets