Learn More
Aalborg™ Needle Valve for Aluminum Flowmeter
Aalborg™ Needle Valve for Aluminum Flowmeter is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
Brand:  Aalborg™ VCL-AB
 View more versions of this product
Product Code. 11882350
  • 118.00€ / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
  • They may also be used as shut off valves.
  • Bodies of the valves are reinforced by structurally rigid metallic shells.
  • Shells are made of anodized aluminum bushings are made of plated brass or 316 stainless steel.
Specifications
Specifications
2400 mL/min.
130mL/min.
Low flow
65°C
Standard Valve