missing translation for 'onlineSavingsMsg'
Learn More

Amentoflavone, Thermo Scientific™

Product Code. 17585565
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 1617-53-4
Quantity 100 mg
Molecular Formula C30 H18 O10
Formula Weight 538.48
Physical Form Powder
Color White to Yellow to Beige
Purity ≥97.5%
InChI Key YUSWMAULDXZHPY-UHFFFAOYSA-N
SMILES OC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=C(O)C(=C2O1)C1=CC(=CC=C1O)C1=CC(=O)C2=C(O)C=C(O)C=C2O1
IUPAC Name 8-[5-(5,7-dihydroxy-4-oxo-4H-chromen-2-yl)-2-hydroxyphenyl]-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
Molecular Weight (g/mol) 538.46
Chemical Name or Material Amentoflavone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.