Learn More
Ammonium cobalt(II) phosphate, anhydrous, 98%, Thermo Scientific Chemicals
44.72€ - 147.74€
Chemical Identifiers
CAS | 14590-13-7 |
---|---|
Molecular Formula | CoH4NO4P |
Molecular Weight (g/mol) | 171.94 |
MDL Number | MFCD02683100 |
InChI Key | FCXHHGTWMFGYPP-UHFFFAOYSA-L |
Synonym | ammonium cobaltous phosphate, ammonium cobalt ii phosphate, ammonium cobalt phosphate, kobaltammoniumphosphat, acmc-20alt0, ammonium cobalt orthophosphate, azanium cobalt 2+ phosphate, phosphoric acid, ammonium cobalt 2+ salt 1:1:1, ammonium cobalt ii phosphate, anhydrous, phosphoric acid,ammonium cobalt 2+ salt 1:1:1 |
PubChem CID | 167120 |
IUPAC Name | azanium;cobalt(2+);phosphate |
SMILES | [NH4+].[Co++].[O-]P([O-])([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429997
|
Thermo Scientific Alfa Aesar
B22934.18 |
50 g |
44.72€
50g |
In Stock
Log in to see stock. |
|||||
11439997
|
Thermo Scientific Alfa Aesar
B22934.30 |
250 g |
147.74€
250g |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14590-13-7 | |
171.94 | |
FCXHHGTWMFGYPP-UHFFFAOYSA-L | |
167120 | |
[NH4+].[Co++].[O-]P([O-])([O-])=O |
CoH4NO4P | |
MFCD02683100 | |
ammonium cobaltous phosphate, ammonium cobalt ii phosphate, ammonium cobalt phosphate, kobaltammoniumphosphat, acmc-20alt0, ammonium cobalt orthophosphate, azanium cobalt 2+ phosphate, phosphoric acid, ammonium cobalt 2+ salt 1:1:1, ammonium cobalt ii phosphate, anhydrous, phosphoric acid,ammonium cobalt 2+ salt 1:1:1 | |
azanium;cobalt(2+);phosphate |
Specifications
14590-13-7 | |
NH4CoPO4 | |
14,513 | |
FCXHHGTWMFGYPP-UHFFFAOYSA-L | |
azanium;cobalt(2+);phosphate | |
167120 | |
98% |
CoH4NO4P | |
MFCD02683100 | |
ammonium cobaltous phosphate, ammonium cobalt ii phosphate, ammonium cobalt phosphate, kobaltammoniumphosphat, acmc-20alt0, ammonium cobalt orthophosphate, azanium cobalt 2+ phosphate, phosphoric acid, ammonium cobalt 2+ salt 1:1:1, ammonium cobalt ii phosphate, anhydrous, phosphoric acid,ammonium cobalt 2+ salt 1:1:1 | |
[NH4+].[Co++].[O-]P([O-])([O-])=O | |
171.94 | |
171.94 | |
Ammonium cobalt(II) phosphate, anhydrous |
Safety and Handling
GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P330-P333+P313-P363-P501c
H302-H317-H350
missing translation for 'einecsNumber' : 238-633-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.