missing translation for 'onlineSavingsMsg'
Learn More
Ammonium hydroxide solution, TraceSELECT Ultra, for ultratrace analysis,≥20% in H2O, Fluka, Honeywell Product Code.: 15651560

Ammonium hydroxide solution, TraceSELECT Ultra, for ultratrace analysis,≥20% in H2O, Fluka, Honeywell

Product Code. 15651560 Shop All Honeywell Fluka Products
1 L
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy

Product Code. 15651560

Brand: Honeywell Fluka 167481L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5, 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
MDL Number MFCD00066650
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ammonia aqueous, Ammonia water
IUPAC Name water
SMILES O

Specifications

Boiling Point 24°C
CAS Min % ≥70.00, ≥20.00
CAS Max % <90.00, <30.00
Beilstein 3587154
Density 0.90 g/cm3
Quantity 1 L
Grade Trace Analysis
Packaging Poly Bottle
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium hydroxide solution
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ammonia, aqueous solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium hydroxide solution, TraceSELECT Ultra, for ultratrace analysis,≥20% in H2O, Fluka, Honeywell >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.