Learn More
Zeiss™ Analyzer Slider
Features 360° rotatable position 1 with analyzer and position 2 with clear aperture to allow mounting of filters with 32mm diameter
Brand: Zeiss™ 428103-0000-000
This item is not returnable.
View return policy
Description
Accessory for Zeiss MicroscopesSpecifications
Analyzer Slider D/A | |
32mm dia.: mounting of filters | |
Analyzer and clear aperture for mounting of filters with 32mm dia. | |
1 Ea. |