Learn More
ABEBA™ Anatom Black/Red Safety Shoes S3 ESD
Red/black Anatom shoes made of water/tear resistant Cordura, slip resistant PU2D outsole, steel toe cap, textile antipuncture
Brand:  ABEBA™ 32238/37
 View more versions of this product
Product Code. 17121205
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
  • Comfortable, robust and durable shoes
  • Comply to EN ISO 20345: 2012, S3, SRC
  • ESD dissipative according to EN ISO 61340
  • Slip resistant PU double density outsole
  • Black/red water and tear resistant Cordura
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Scratch-resistant coated fabric in toe and heel area
  • Breathable and antibacterial lining with silver ions
  • Removable active comfort insole
Specifications
Specifications
Black, Red
37
EN ISO 20345: 2012 S3 SRC, EN ISO 61340 ESD class 1
Low-cut
S3 SRC ESD
Cordura
Steel
Men's
Water Resistant