Learn More
anti-ALPL, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00000249-PW2.1Set
Includes: Antibody pair for IP: rabbit purified polyclonal anti-ALPL (300μg), Antibody pair for WB: mouse purified polyclonal anti-ALPL (50μg)
Specifications
ALPL | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
AP-TNAP/FLJ40094/FLJ93059/HOPS/MGC161443/MGC167935/TNAP/TNSALP | |
Mouse, Rabbit | |
RUO | |
249 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
ALPL | |
ALPL | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: rabbit purified polyclonal anti-ALPL (300 ug) 2. Antibody pair for WB: mouse purified polyclonal anti-ALPL (50 ug) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only