Learn More
anti-ALPL, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00000249-PW2.1Set
Product Code. 16171714
  • 809.00€ / 1 set
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Antibody pair for IP: rabbit purified polyclonal anti-ALPL (300μg), Antibody pair for WB: mouse purified polyclonal anti-ALPL (50μg)
Specifications
ALPL
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
AP-TNAP/FLJ40094/FLJ93059/HOPS/MGC161443/MGC167935/TNAP/TNSALP
Mouse, Rabbit
RUO
249
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
ALPL
ALPL
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit purified polyclonal anti-ALPL (300 ug) 2. Antibody pair for WB: mouse purified polyclonal anti-ALPL (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.