Learn More
anti-ANXA4 Matched Antibody Pair, Abnova
Brand: Abnova H00000307-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-ANXA4 (100μg); Mouse purified polyclonal anti-ANXA4 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells.
ELISA
Specifications
ANXA4 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ANXA4. | |
ANX4/DKFZp686H02120/MGC75105/PIG28/ZAP36 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-ANXA4 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-ANXA4 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
ANXA4 | |
ANXA4 | |
Affinity Purified | |
RUO | |
307 | |
Human |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.