missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-ARHGDIA Matched Antibody Pair, Abnova
Brand: Abnova H00000396-AP11.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-ARHGDIA (100μg); Mouse monoclonal anti-ARHGDIA, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Aplysia Ras-related homologs (ARHs), also called Rho genes, belong to the RAS gene superfamily encoding small guanine nucleotide exchange (GTP/GDP) factors. The ARH proteins may be kept in the inactive, GDP-bound state by interaction with GDP dissociation inhibitors, such as ARHGDIA.
ELISA
Specifications
ARHGDIA | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ARHGDIA. | |
GDIA1/MGC117248/RHOGDI/RHOGDI-1 | |
Mouse, Rabbit | |
1 Set | |
Signal Transduction | |
396 | |
Human | |
IgG1 |
ELISA | |
Unconjugated | |
ARHGDIA | |
ARHGDIA | |
Affinity Purified | |
RUO | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-ARHGDIA (100 μg) 2. Detection antibody: mouse monoclonal anti-ARHGDIA, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |