Learn More
anti-CBFA2T2, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00009139-AP41.1Set
Product Code. 16152974
  • 809.00€ / 1 set
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Capture antibody: mouse monoclonal anti-CBFA2T2, IgG2a Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-CBFA2T2, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
CBFA2T2
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CBFA2T2.
DKFZp313F2116/EHT/MTGR1/ZMYND3
Mouse
RUO
9139
Human
Liquid
ELISA
Biotin
CBFA2T2
CBFA2T2
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CBFA2T2, IgG2a Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CBFA2T2, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.