Learn More
CD274 Mouse anti-Human, Brilliant Violet 421, Clone: MIH1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 563738
414.39 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The MIH1 monoclonal antibody specifically binds to CD274, also known as Programmed Death Ligand 1 (PD-L1) and B7 Homolog 1 (B7-H1). CD274/PD-L1 and CD273/PD-L2 are members of the B7 family within the Immunoglobulin superfamily. These type I transmembrane glycoproteins function as ligands for CD279, likewise known as the Programmed Death 1 (PD-1) receptor. CD274 is expressed on activated dendritic cells, macrophages and T cells. Studies show some overlapping functions of CD274 and CD273 and indicate an important role for CD279 and its ligands in regulating T and B cell responses. CD274 is also expressed on placental trophoblasts, myocardial endothelium, cortical thymic epithelial cells, keratinocytes, and carcinomas. The MIH1 antibody is reportedly a blocking antibody.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.

Specifications
CD274 | |
Monoclonal | |
Brilliant Violet 421 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human B7-H1 Transfected Cell Line | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
MIH1 | |
B7-H1; Programmed death ligand 1; PD-L1; PDCD1 ligand 1; PDCD1L1; PDCD1LG1 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.