missing translation for 'onlineSavingsMsg'
Learn More

anti-CD40LG Matched Antibody Pair, Abnova™

Brand:  Abnova H00000959-AP41.1Set

Product Code. 16181864

  • 809.00€ / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Capture antibody: mouse monoclonal anti-CD40LG (100μg); Detection antibody: biotinylated mouse monoclonal anti-CD40LG, IgG1 Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
This item is not returnable. View return policy

Description

Description

Host Species: Mouse, Mouse
Isotype: detection antibody: IgG1, κ
Cross Reactivity: Human

ELISA Pair

Specifications

Specifications

CD40LG
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CD40LG.
CD154/CD40L/HIGM1/IGM/IMD3/T-BAM/TNFSF5/TRAP/gp39/hCD40L
Mouse
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CD40LG, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CD40LG, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
Biotin
CD40LG
CD40LG
1 Set
959
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
anti-CD40LG Matched Antibody Pair, Abnova™ > 1 Set; Unlabeled, Biotin

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.