missing translation for 'onlineSavingsMsg'
Learn More

CtBP1 (C-terminal Binding Protein-1) Mouse, Unlabeled, Clone: 3, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 612042

180.33 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15876079

  • 181.00€ / 1 set

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Transcriptional repression is critical for genetic regulation of many cellular proteins, and involves either a transcription regulator that has an intrinsic repressor domain, or a transcription regulator that contains a binding site in the regulatory domain where a corepressor can bind. C-terminal binding proteins (CtBP1 and CtBP2) are a class of corepressors that interact with a subset of transcription factors through a PLDLSL sequence motif. CtBP1 and CtBP2 are 80% homologous, and both are expressed at high levels in embryonic tissues. However, CtBP1 is expressed higher in adult tissues, and is more widely expressed in embryonic and adult tissues. Both CtBP1 and CtBP2 bind the δEF1 zinc finger-homeodomain transcription factor, and enhance transcriptional repression via interaction with the PLDLSL-sequence in δEF1. CtBP1 and CtBP2 bind other zinc finger transcription factors, including Kheper and BKLF. In addition, CtBP2 interacts with other transcription factors, such as hFOG-2, Evi-1, AREB6, and ZEB. Thus, CtBP1 and CtBP2 may be important corepressors for a variety of transcriptional factors.

Host Species: Mouse
Clone: 3
Isotype: IgG1
Species Reactivity [for Features Main]: Mouse
Immunogen: Mouse CtBP1 aa. 345-441

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CtBP1 (C-terminal Binding Protein-1)
Monoclonal
250μg/mL
C-terminal Binding Protein-1
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
3
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse CtBP1 aa. 345-441
50 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CtBP1 (C-terminal Binding Protein-1) Mouse, Unlabeled, Clone: 3, BD > 50μg; Unlabeled

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.