missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-EDN1 Matched Antibody Pair, Abnova™
Brand: Abnova H00001906-AP51.1Set
Includes: Capture antibody: mouse monoclonal anti-EDN1, IgG2a κ (100μg); Detection antibody: rabbit purified polyclonal anti-EDN1 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human EDN1
Host Species: Mouse, Rabbit
Isotype: IgG2a κ, IgG
Species Reactivity: Hu
ELISA Pair (Transfected Lysate)
Specifications
EDN1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human EDN1. | |
ET1/HDLCQ7 | |
Mouse, Rabbit | |
RUO | |
1906 | |
Human | |
Purified |
ELISA | |
Unconjugated | |
EDN1 | |
EDN1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-EDN1, IgG2a Kappa (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-EDN1 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
IgG, IgG2a κ |