Learn More
FTL, Human, Matched Antibody Pair (AP48), Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00002512-AP48.1Set
Product Code. 16182064
  • 809.00€ / 1 set
Includes: Capture antibody: mouse monoclonal anti-FTL, IgG1 Kappa (100ÎĽg) , Detection antibody: biotinylated mouse monoclonal anti-FTL, IgG1 Kappa (50ÎĽg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
FTL
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FTL.
MGC71996
Mouse
RUO
2512
Human
Liquid
ELISA
Biotin
FTL
FTL
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-FTL, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-FTL, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
FTL, Human, Matched Antibody Pair (AP48), Abnova™ > 1 Set