missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-FXN Matched Antibody Pair, Abnova
Brand: Abnova H00002395-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-FXN (100μg); Mouse purified polyclonal anti-FXN (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This nuclear gene encodes a mitochondrial protein which belongs to FRATAXIN family. The protein functions in regulating mitochondrial iron transport and respiration. The expansion of intronic trinucleotide repeat GAA results in Friedreich ataxia. Alternative splicing results in multiple transcript variants.
ELISA
Specifications
FXN | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FXN. | |
CyaY/FA/FARR/FRDA/MGC57199/X25 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FXN (100 μg) 2. Detection antibody: mouse purified polyclonal anti-FXN (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
FXN | |
FXN | |
Affinity Purified | |
RUO | |
2395 | |
Human |