missing translation for 'onlineSavingsMsg'
Learn More

Glutamine Synthetase Mouse, Unlabeled, Clone: 6, BD

Mouse Monoclonal Antibody

216.00€ - 343.00€

Specifications

Antigen Glutamine Synthetase
Clone 6
Concentration 250ÎĽg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15855499
View Documents
BD Biosciences
610517
50 ÎĽg
216.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
15865499
View Documents
BD Biosciences
610518
150 ÎĽg
343.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Glutamine synthetase catalyzes the amination of glutamic acid to form glutamine. It is found in mammals as an octamer of identical 45 kDa subunits. Glutamine synthetase activity is a useful marker for astrocytes and an important differentiation feature in retina. It is also considered to be a key enzyme in the recycling of the neurotransmitter glutamate.

Specifications

Specifications

Glutamine Synthetase
250ÎĽg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG2a
Affinity Purified
Glutamate-Ammonia Ligase; GLUL; GLNS
6
Western Blot
Unconjugated
RUO
Human, Mouse, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glutamine Synthetase Mouse, Unlabeled, Clone: 6, BD

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.