Learn More
HMGB2, Human, Matched Antibody Pair (AP45), Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00003148-AP45.1Set
Product Code. 16192164
  • 809.00€ / 1 set
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Includes: Capture antibody: mouse monoclonal anti-HMGB2 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-HMGB2, IgG Mix Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
HMGB2
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HMGB2.
HMG2
Mouse
RUO
3148
Human
Liquid
ELISA
Biotin
HMGB2
HMGB2
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-HMGB2 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-HMGB2, IgG Mix Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
HMGB2, Human, Matched Antibody Pair (AP45), Abnova™ > 1 Set