missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-HMGCL Matched Antibody Pair, Abnova™
Brand: Abnova H00003155-AP51.1Set
Includes: Capture antibody: mouse monoclonal anti-HMGCL (100μg); Detection antibody: rabbit purified polyclonal anti-HMGCL (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Host Species: Mouse, Rabbit
Cross Reactivity: Human
ELISA Pair
Specifications
HMGCL | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HMGCL. | |
HL | |
Mouse, Rabbit | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-HMGCL (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-HMGCL (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
HMGCL | |
HMGCL | |
1 Set | |
3155 | |
Human |