missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Ig, PE-CF594, Clone: R35-95, BD
Brand: BD Biosciences 562302
352.42 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Host Species: Rat
Clone: R35-95
Isotype: IgG2a, Îş
Immunogen: Mouse pooled immunoglobulin
Flow Cytometry, Isotype Control

Specifications
Flow Cytometry, Immunocytology | |
0.2mg/mL | |
Rat | |
Affinity Purified | |
4°C |
R35-95 | |
PE-CF594 | |
Mouse pooled immunoglobulin | |
0.1 mg | |
IgG2a Îş |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
anti-Ig, PE-CF594, Clone: R35-95, BD > 0.1mg; PE-CF594
Spot an opportunity for improvement?Share a Content Correction