Learn More
anti-IgG1, Îş, R-PE, Clone: MOPC-21, BD
Brand: BD Biosciences 559320
267.25 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody
development only or reported in the literature.
Flow Cytometry, Isotype Control

Specifications
Flow Cytometry | |
R-PE | |
Affinity Purified | |
Undiluted at 4°C |
MOPC-21 | |
Mouse | |
100 Tests | |
IgG1 Îş |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.