Learn More
anti- IgG2a κ, Mouse, BV421, Clone: G155-178, BD
Isotype Control
Brand: BD Biosciences 562439
Description
The G155-178 clone has an unknown specificity. Trinitrophenol (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to immunoglobulin Fc receptors. The immunoglobulin secreted by the G155-178 hybridoma was selected as an mouse IgG2a, ? isotype control following screening for low background binding on a variety of mouse and human tissues. The antibody was conjugated to Brilliant Violet 421 and has been developed in collaboration with Sirigen. With an Ex Max of 407-nm and Em Max at 421-nm, Brilliant Violet 421 can be excited by the violet laser and detected in the standard Pacific Blue filter set (eg,
450/50-nm filter). Brilliant Violet 421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a, κ
Immunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control

Specifications
Flow Cytometry, Immunocytology | |
Brilliant Violet 421 | |
TNP-keyhole limpet hemocyanin | |
50 μg | |
IgG2a κ |
G155-178 | |
Mouse | |
Affinity Purified | |
4°C, protect from prolonged exposure to light, DO NOT FREEZE |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.