Learn More
anti-IgG2b, Îş, FITC, Clone: MPC-11, BD
Brand: BD Biosciences 559532
219.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The MPC-11 antibody has unknown specificity. The MPC-11 cell line was adapted to continuous culture from the tranplantable Merwin Plasmacytoma-11, which was induced by intraperitoneal implantation of a Millipore diffusion chamber into a BALB/c mouse. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry, Isotype Control

Specifications
Flow Cytometry | |
0.5mg/mL | |
Mouse | |
0.25 mg | |
IgG2b Îş |
MPC-11 | |
FITC | |
Affinity Purified | |
Undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.