missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2b κ, Mouse , Clone: MPC-11, BD

Brand:  BD Biosciences 557351

174.93 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15858888

  • 175.00€ / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The MPC-11 antibody has unknown specificity. The MPC-11 cell line was adapted to continuous culture from the tranplantable Merwin Plasmacytoma-11, which was induced by intraperitoneal implantation of a Millipore diffusion chamber into a BALB/c mouse. This antibody is routinely tested by ELISA, immunohistochemical and flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: MPC-11
Isotype: IgG2b, κ

Cytotoxicity Assay, ELISA Standard, Flow Cytometry, Immunohistochemistry (Formalin-Antigen Retrieval Required), Immunohistochemistry (Frozen), Isotype Control

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytotoxicity Assay, ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
0.5mg/mL
Mouse
0.5 mg/mL
IgG2b κ
MPC-11
Unconjugated
Affinity Purified
Undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
anti-IgG2b κ, Mouse , Clone: MPC-11, BD > 0.5mg; Unlabeled

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.