missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2b, Îş Mouse, R-PE, Clone: 27-35, BD

Brand:  BD Biosciences 555058

296.95 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15807988

  • 297.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The mouse IgG2b, Îş immunoglobulin isotype control monoclonal antibody 27-35 is specific for the hapten dansyl (5-[dimethylamino] naphthalene-1-sulfonyl). This hapten is not expressed on human cells or human cell lines. The 27-35 immunoglobulin was selected as an isotype control following testing which demonstrated low background staining on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow Cytometry
0.2mg/mL
Mouse
0.1 mg
IgG2b Îş
27-35
R-PE
Affinity Purified
Undiluted at 4°C. Protect from exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
anti-IgG2b, Îş Mouse, R-PE, Clone: 27-35, BD > 0.1mg; R-PE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.