missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgM, κ, Clone: R4-22, BD
Brand: BD Biosciences 562489
This item is not returnable.
View return policy
Description
The R4-22 antibody has unknown specificity. This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488nm), green (532nm) and yellow-green (561nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20nm filter).
Host Species: Rat
Clone: R4-22
Isotype: IgM, κ
Immunogen: Pooled Mouse Ig
Flow Cytometry, Isotype Control
Specifications
IgM, κ | |
R4-22 | |
PE-CF594 | |
Pooled Mouse Ig | |
100 μg | |
Primary |
Monoclonal | |
0.2mg/mL | |
Rat | |
Affinity Purified | |
RUO | |
IgM κ |