Learn More
Ki-67 Mouse, V450, Clone: B56, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561281
319.63 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The B56 monoclonal antibody specifically binds to the Ki-67 antigen that is expressed in the nucleus of cycling cells (G1, S, G2, M cell cycle phases). During the G0 phase, the antigen cannot be detected. During interphase of the cell cycle, it is associated with nucleolar components, and it is on the surface of the chromosomes during M phase. Ki-67 is a large protein having 2 alternatively spliced isoforms, an N-terminal forkhead-associated domain, a C-terminal domain that binds to heterochromatin proteins, and multiple phosphorylation sites, the functions of which are still unclear. Because of the strict association of Ki-67 expression with cell proliferation, anti-Ki-67 antibodies are useful for the identification, quantification, and monitoring of growing cell populations.
The antibody is conjugated to BD Horizon V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450 nm. Conjugates with BD Horizon V450 can be used in place of Pacific Blue™ conjugates.

Specifications
Ki-67 | |
Monoclonal | |
Violet Fluor 450 (V450) | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Human Ki-67 | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
B56 | |
MKI67; Antigen identified by monoclonal antibody Ki-67; KIA | |
Mouse | |
Affinity Purified | |
RUO | |
Human, Mouse, Rat, Rhesus Monkey | |
IgG1 Îş |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.