missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-LIPG Ab-1 Matched Antibody Pair, Abnova™
Brand: Abnova H00009388-AP11.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-LIPG (100μg); Detection antibody: mouse monoclonal anti-LIPG, IgG2a Kappa (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human LIPG
Host Species: Rabbit, Mouse
Isotype: IgG, IgG2a Îş
Species Reactivity: Hu
ELISA Pair
Specifications
LIPG | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human LIPG. | |
EDL/EL/PRO719 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-LIPG (100 μg) 2. Detection antibody: mouse monoclonal anti-LIPG, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
LIPG | |
LIPG | |
Affinity Purified | |
RUO | |
9388 | |
Human | |
IgG, IgG2a Îş |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
anti-LIPG Ab-1 Matched Antibody Pair, Abnova™ > 1 Set, Unlabeled
Spot an opportunity for improvement?Share a Content Correction