Learn More
anti-MATN2, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00004147-PW1.1Set
Product Code. 16026728
  • 809.00€ / 1 set
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Includes: Antibody pair for IP: rabbit polyclonal anti-MATN2 (300μL), Antibody pair for WB: mouse purified polyclonal anti-MATN2 (50μg)
Specifications
MATN2
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
MATN2
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-MATN2 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-MATN2 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Immunoprecipitation, Western Blot
Unconjugated
MATN2
Mouse, Rabbit
RUO
4147