Learn More
MMP3, Human, IP-WB Antibody Pair (PW1), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00004314-PW1.1Set
Product Code. 16086958
  • 809.00€ / 1 set
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Antibody pair for IP: rabbit polyclonal anti-MMP3 (300μL), Antibody pair for WB: mouse purified polyclonal anti-MMP3 (50μg)
Specifications
MMP3
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
CHDS6/MGC126102/MGC126103/MGC126104/MMP-3/SL-1/STMY/STMY1/STR1
Mouse, Rabbit
RUO
4314
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
MMP3
MMP3
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-MMP3 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-MMP3 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.