missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-NELL1 Matched Antibody Pair, Abnova™
Brand: Abnova H00004745-AP51.1Set
Includes: Capture antibody: mouse monoclonal anti-NELL1 (100μg); Detection antibody: rabbit purified polyclonal anti-NELL1 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NELL1.
Host Species: Mouse, Rabbit
Species Reactivity: Hu
ELISA Pair (Transfected Lysate)
Specifications
NELL1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NELL1. | |
FLJ45906/IDH3GL/NRP1 | |
Mouse, Rabbit | |
RUO | |
4745 | |
Human | |
Purified |
ELISA | |
Unconjugated | |
NELL1 | |
NELL1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-NELL1 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-NELL1 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
anti-NELL1 Matched Antibody Pair, Abnova™ > 1 set, Unlabeled
Spot an opportunity for improvement?Share a Content Correction