missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-OPTN Matched Antibody Pair, Abnova™
Brand: Abnova H00010133-AP21.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-OPTN (100μg); Detection antibody: mouse purified polyclonal anti-OPTN (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human OPTN
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair
Specifications
OPTN | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human OPTN. | |
FIP2/GLC1E/HIP7/HYPL/NRP/TFIIIA-INTP | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-OPTN (100 μg) 2. Detection antibody: mouse purified polyclonal anti-OPTN (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
OPTN | |
OPTN | |
Affinity Purified | |
RUO | |
10133 | |
Human |