Learn More
anti-Ornithine Carbamoyltransferase, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NBP1-31582
Additional Details : Weight : 0.00970kg
Product Code. 15715847
  • 751.50€ / 100µL
Estimated Shipment: 04-07-2024
to see stock.
Añadir a la cesta
Descripción
Descripción
Ornithine Carbamoyltransferase Polyclonal specifically detects Ornithine Carbamoyltransferase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Ofertas especiales
Ofertas especiales
For Research Use Only