missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-OXSR1 Matched Antibody Pair, Ab-1, Abnova
Brand: Abnova H00009943-AP11.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-OXSR1 (100μg); Mouse monoclonal anti-OXSR1, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
The product of this gene belongs to the Ser/Thr protein kinase family of proteins. It regulates downstream kinases in response to environmental stress, and may play a role in regulating the actin cytoskeleton.
ELISA
Specifications
OXSR1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human OXSR1. | |
KIAA1101/OSR1 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-OXSR1 (100 μg) 2. Detection antibody: mouse monoclonal anti-OXSR1, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
OXSR1 | |
OXSR1 | |
Affinity Purified | |
RUO | |
9943 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
anti-OXSR1 Matched Antibody Pair, Ab-1, Abnova > 1 Set
Spot an opportunity for improvement?Share a Content Correction