missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMT, Human, IP-WB Antibody Pair (PW2), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00005409-PW2.1Set
Includes: Antibody pair for IP: mouse monoclonal anti-PNMT (300μg), Antibody pair for WB: rabbit polyclonal anti-PNMT (50μL)
Specifications
PNMT | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
MGC34570/PENT/PNMTase | |
Mouse, Rabbit | |
RUO | |
5409 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
PNMT | |
PNMT | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-PNMT (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-PNMT (50 ul) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only