Learn More
PSMA7, Human, IP-WB Antibody Pair (PW2), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00005688-PW2.1Set
Product Code. 16038418
  • 809.00€ / 1 set
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Antibody pair for IP: rabbit polyclonal anti-PSMA7 (300μL), Antibody pair for WB: mouse polyclonal anti-PSMA7 (50μL)
Specifications
PSMA7
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
C6/HSPC/MGC3755/RC6-1/XAPC7
Mouse, Rabbit
RUO
5688
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
PSMA7
PSMA7
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-PSMA7 (300 ul) 2. Antibody pair for WB: mouse polyclonal anti-PSMA7 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.